Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTCCTAGGTCTTTAATGTGGG[C/G]TTTGAGTTAATCATTAAGAGTTAGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607467 | |||||||||||||||||||||||
Literature Links: |
CLECL1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CLECL1 - C-type lectin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253750.1 | Intron | NP_001240679.1 | ||||
NM_001267701.1 | Intron | NP_001254630.1 | ||||
NM_172004.3 | Intron | NP_742001.1 | ||||
XM_011520574.2 | Intron | XP_011518876.1 | ||||
XM_017018885.1 | Intron | XP_016874374.1 |
Set Membership: |
HapMap |