Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCGTTTCTTCTTTTTAATTTCA[A/G]TACAACTGTTTACTCACCTGGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608301 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LGI2 - leucine rich repeat LGI family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018176.3 | Intron | NP_060646.2 | ||||
XM_011513850.2 | Intron | XP_011512152.1 | ||||
XM_017008356.1 | Intron | XP_016863845.1 |