Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137241 MIM: 601061 | ||||||||||||||||||||||||||||||||
Literature Links: |
GIPR PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GIPR - gastric inhibitory polypeptide receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
QPCTL - glutaminyl-peptide cyclotransferase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163377.1 | Intron | NP_001156849.1 | ||||
NM_017659.3 | Intron | NP_060129.2 | ||||
XM_011527048.2 | Intron | XP_011525350.1 | ||||
XM_017026900.1 | Intron | XP_016882389.1 |
SNRPD2 - small nuclear ribonucleoprotein D2 polypeptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |