Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605987 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6736 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MIR6736 - microRNA 6736 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT17 - nudix hydrolase 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012758.2 | 568 | Intron | NP_001012776.1 | |||
XM_011509260.2 | 568 | UTR 5 | XP_011507562.1 | |||
XM_011509262.1 | 568 | UTR 5 | XP_011507564.1 | |||
XM_017000560.1 | 568 | UTR 5 | XP_016856049.1 |
PIAS3 - protein inhibitor of activated STAT 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3C - RNA polymerase III subunit C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |