Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAACAACAAAGTCCACTTTTCTT[C/G]TTTTGCTATAATAGATATGCTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602659 MIM: 616518 MIM: 611023 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MNAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MNAT1 - MNAT1, CDK activating kinase assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC38A6 - solute carrier family 38 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT5 - tRNA methyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020810.3 | 2615 | UTR 3 | NP_065861.3 | |||
XM_005267916.3 | 2615 | UTR 3 | XP_005267973.2 | |||
XM_011537017.2 | 2615 | Intron | XP_011535319.1 |