Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCCTTCCCTGTCACTAGTGTCC[C/T]TGCTTCCTCCCCTACCCTGGTTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604087 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP46A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CYP46A1 - cytochrome P450 family 46 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006668.1 | Intron | NP_006659.1 | ||||
XM_005267274.4 | Intron | XP_005267331.1 | ||||
XM_011536364.1 | Intron | XP_011534666.1 | ||||
XM_011536365.1 | Intron | XP_011534667.1 | ||||
XM_017020933.1 | Intron | XP_016876422.1 |
HHIPL1 - HHIP like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |