Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATAATTCTAAAACTTCACTTTTG[A/T]CCTTAAATCAATATATTACGGATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601366 | ||||||||||||||||||||
Literature Links: |
SMAD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMAD2 - SMAD family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003652.3 | 7753 | UTR 3 | NP_001003652.1 | |||
NM_001135937.2 | 7753 | UTR 3 | NP_001129409.1 | |||
NM_005901.5 | 7753 | UTR 3 | NP_005892.1 | |||
XM_005258259.3 | 7753 | UTR 3 | XP_005258316.1 | |||
XM_006722451.3 | 7753 | UTR 3 | XP_006722514.1 | |||
XM_011525984.2 | 7753 | UTR 3 | XP_011524286.1 | |||
XM_011525985.2 | 7753 | UTR 3 | XP_011524287.1 | |||
XM_017025745.1 | 7753 | UTR 3 | XP_016881234.1 | |||
XM_017025746.1 | 7753 | UTR 3 | XP_016881235.1 | |||
XM_017025747.1 | 7753 | UTR 3 | XP_016881236.1 | |||
XM_017025748.1 | 7753 | UTR 3 | XP_016881237.1 | |||
XM_017025749.1 | 7753 | Intron | XP_016881238.1 | |||
XM_017025750.1 | 7753 | UTR 3 | XP_016881239.1 |