Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAACAGAATTTTTGTTAAAATA[C/T]GTATCGATACTGAAAATAAATCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611692 | ||||||||||||||||||||
Literature Links: |
ZBTB34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZBTB34 - zinc finger and BTB domain containing 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099270.1 | Intron | NP_001092740.1 | ||||
XM_005251989.4 | Intron | XP_005252046.1 | ||||
XM_011518699.2 | Intron | XP_011517001.1 |