Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAATGGAATCAAAGCATTTGCC[A/G]CTGAATGAGAGTTGGTGTGTGAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603109 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723493 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102723493 - uncharacterized LOC102723493 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMAD3 - SMAD family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145102.1 | Intron | NP_001138574.1 | ||||
NM_001145103.1 | Intron | NP_001138575.1 | ||||
NM_001145104.1 | Intron | NP_001138576.1 | ||||
NM_005902.3 | Intron | NP_005893.1 | ||||
XM_011521559.2 | Intron | XP_011519861.1 | ||||
XM_011521560.1 | Intron | XP_011519862.1 |
Set Membership: |
HapMap |