Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGAAATTAATTTGCTAACAGACA[C/G]AAAACTTGTCAACATCCACAGAGTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 132810 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHX1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EPHX1 - epoxide hydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000120.3 | Intron | NP_000111.1 | ||||
NM_001136018.3 | Intron | NP_001129490.1 | ||||
NM_001291163.1 | Intron | NP_001278092.1 |
TMEM63A - transmembrane protein 63A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014698.2 | Intron | NP_055513.2 | ||||
XM_006711841.3 | Intron | XP_006711904.1 | ||||
XM_011544328.2 | Intron | XP_011542630.1 | ||||
XM_011544329.2 | Intron | XP_011542631.1 | ||||
XM_011544330.2 | Intron | XP_011542632.1 | ||||
XM_011544331.2 | Intron | XP_011542633.1 | ||||
XM_011544332.2 | Intron | XP_011542634.1 | ||||
XM_017002916.1 | Intron | XP_016858405.1 |
Set Membership: |
HapMap |