Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136351 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLT3 - fms related tyrosine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004119.2 | 3565 | UTR 3 | NP_004110.2 | |||
XM_011535015.2 | 3565 | Intron | XP_011533317.1 | |||
XM_011535017.2 | 3565 | Intron | XP_011533319.1 | |||
XM_011535018.2 | 3565 | Intron | XP_011533320.1 | |||
XM_017020486.1 | 3565 | Intron | XP_016875975.1 | |||
XM_017020487.1 | 3565 | Intron | XP_016875976.1 | |||
XM_017020488.1 | 3565 | Intron | XP_016875977.1 | |||
XM_017020489.1 | 3565 | Intron | XP_016875978.1 |
Set Membership: |
HapMap |