Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137162 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GABRR2 - gamma-aminobutyric acid type A receptor rho2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002043.4 | 4629 | UTR 3 | NP_002034.3 | |||
XM_011535713.2 | 4629 | Intron | XP_011534015.1 | |||
XM_011535714.2 | 4629 | Intron | XP_011534016.1 | |||
XM_011535715.2 | 4629 | Intron | XP_011534017.1 | |||
XM_011535716.2 | 4629 | Intron | XP_011534018.1 | |||
XM_011535717.2 | 4629 | Intron | XP_011534019.1 |
Set Membership: |
HapMap Validated |