Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609787 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC114224 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC114224 - uncharacterized LOC114224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBAP1 - ubiquitin associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171201.1 | Intron | NP_001164672.1 | ||||
NM_001171202.1 | Intron | NP_001164673.1 | ||||
NM_001171203.2 | Intron | NP_001164674.1 | ||||
NM_001171204.2 | Intron | NP_001164675.1 | ||||
NM_016525.4 | Intron | NP_057609.2 | ||||
XM_006716779.3 | Intron | XP_006716842.1 | ||||
XM_011517898.2 | Intron | XP_011516200.1 | ||||
XM_011517899.1 | Intron | XP_011516201.1 | ||||
XM_011517900.1 | Intron | XP_011516202.1 | ||||
XM_017014801.1 | Intron | XP_016870290.1 | ||||
XM_017014802.1 | Intron | XP_016870291.1 |