Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGAGGGATCTTGATAGGGTTCCA[A/T]CAGTTTCACTTTCCTTTCCCCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604650 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFIT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFIT3 - interferon induced protein with tetratricopeptide repeats 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031683.3 | Intron | NP_001026853.1 | ||||
NM_001289758.1 | Intron | NP_001276687.1 | ||||
NM_001289759.1 | Intron | NP_001276688.1 | ||||
NM_001549.5 | Intron | NP_001540.2 |