Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAATCTTGGAACCTCATATTCGA[C/T]AGGCACGGCTGTAAGATTGAGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603322 MIM: 609507 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDUFB6 - NADH:ubiquinone oxidoreductase subunit B6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199987.1 | Intron | NP_001186916.1 | ||||
NM_002493.4 | Intron | NP_002484.1 | ||||
NM_182739.2 | Intron | NP_877416.1 |
TOPORS - TOP1 binding arginine/serine rich protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOPORS-AS1 - TOPORS antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |