Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600046 MIM: 608872 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ABCA1 - ATP binding cassette subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005502.3 | 9001 | UTR 3 | NP_005493.2 | |||
XM_005251773.2 | 9001 | UTR 3 | XP_005251830.1 | |||
XM_005251776.2 | 9001 | UTR 3 | XP_005251833.1 | |||
XM_011518339.2 | 9001 | UTR 3 | XP_011516641.1 | |||
XM_011518340.2 | 9001 | UTR 3 | XP_011516642.1 | |||
XM_011518341.2 | 9001 | UTR 3 | XP_011516643.1 | |||
XM_011518342.2 | 9001 | UTR 3 | XP_011516644.1 | |||
XM_011518344.2 | 9001 | Intron | XP_011516646.1 | |||
XM_017014378.1 | 9001 | UTR 3 | XP_016869867.1 | |||
XM_017014379.1 | 9001 | UTR 3 | XP_016869868.1 | |||
XM_017014380.1 | 9001 | UTR 3 | XP_016869869.1 | |||
XM_017014381.1 | 9001 | UTR 3 | XP_016869870.1 | |||
XM_017014382.1 | 9001 | UTR 3 | XP_016869871.1 |
NIPSNAP3B - nipsnap homolog 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |