Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGAATGGCTTTAGCTATTGTT[C/G]AAAACAGATAATCAGGTTATAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 105850 MIM: 601030 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANG - angiogenin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097577.2 | Intron | NP_001091046.1 | ||||
NM_001145.4 | Intron | NP_001136.1 |
RNASE4 - ribonuclease A family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282192.1 | Intron | NP_001269121.1 | ||||
NM_001282193.1 | Intron | NP_001269122.1 | ||||
NM_002937.4 | Intron | NP_002928.1 | ||||
NM_194431.2 | Intron | NP_919412.1 |