Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTCCTGGGTTACTTCTTGATTT[G/C]CTGCCAGTGGCATATAATGATGACT
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602431 | |||||||||||||||||||||||||||||||||||
Literature Links: |
ROBO2 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |