Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGGCTCCTGTCGTCCGTTCTCT[C/G]AGGCTCCCGTTCAGGTAAAGGGCTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612480 | |||||||||||||||||||||||
Literature Links: |
TIPARP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TIPARP - TCDD inducible poly(ADP-ribose) polymerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184717.1 | 367 | Intron | NP_001171646.1 | |||
NM_001184718.1 | 367 | Intron | NP_001171647.1 | |||
NM_015508.4 | 367 | UTR 5 | NP_056323.2 |
TIPARP-AS1 - TIPARP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |