Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAGGTGTGGTGGATAGGAAACC[A/T]CTTGGGAATCAGTATGCTATGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR70 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WDR70 - WD repeat domain 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018034.2 | Intron | NP_060504.1 | ||||
XM_005248318.3 | Intron | XP_005248375.1 | ||||
XM_011514061.2 | Intron | XP_011512363.1 | ||||
XM_017009604.1 | Intron | XP_016865093.1 | ||||
XM_017009605.1 | Intron | XP_016865094.1 |
Set Membership: |
HapMap |