Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGGCCACCGAGAACTTGTGCAA[A/G]GTGGGCCGGGCCGGGTCGGTCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603958 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100287944 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100287944 - uncharacterized LOC100287944 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFX4 - regulatory factor X4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206691.1 | Intron | NP_001193620.1 | ||||
NM_032491.5 | Intron | NP_115880.2 | ||||
NM_213594.2 | Intron | NP_998759.1 |