Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCCCCGCAGAGCCTCTGACTG[A/G]GCAGGTGGCGGCCCAGGAACCTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615393 MIM: 607505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTERF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTERF4 - mitochondrial transcription termination factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PASK - PAS domain containing serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252119.1 | Intron | NP_001239048.1 | ||||
NM_001252120.1 | Intron | NP_001239049.1 | ||||
NM_001252122.1 | Intron | NP_001239051.1 | ||||
NM_001252124.1 | Intron | NP_001239053.1 | ||||
NM_015148.3 | Intron | NP_055963.2 | ||||
XM_005246991.1 | Intron | XP_005247048.1 | ||||
XM_011510826.1 | Intron | XP_011509128.1 | ||||
XM_011510828.1 | Intron | XP_011509130.1 | ||||
XM_011510829.1 | Intron | XP_011509131.1 | ||||
XM_011510830.1 | Intron | XP_011509132.1 | ||||
XM_011510831.2 | Intron | XP_011509133.1 | ||||
XM_011510832.2 | Intron | XP_011509134.1 | ||||
XM_011510833.1 | Intron | XP_011509135.1 | ||||
XM_011510834.1 | Intron | XP_011509136.1 | ||||
XM_011510835.1 | Intron | XP_011509137.1 | ||||
XM_017003634.1 | Intron | XP_016859123.1 | ||||
XM_017003635.1 | Intron | XP_016859124.1 | ||||
XM_017003636.1 | Intron | XP_016859125.1 |