Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142370 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HCK - HCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM9SF4 - transmembrane 9 superfamily member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014742.3 | Intron | NP_055557.2 | ||||
XM_005260622.4 | Intron | XP_005260679.1 | ||||
XM_017028154.1 | Intron | XP_016883643.1 | ||||
XM_017028155.1 | Intron | XP_016883644.1 | ||||
XM_017028156.1 | Intron | XP_016883645.1 | ||||
XM_017028157.1 | Intron | XP_016883646.1 | ||||
XM_017028158.1 | Intron | XP_016883647.1 | ||||
XM_017028159.1 | Intron | XP_016883648.1 | ||||
XM_017028160.1 | Intron | XP_016883649.1 | ||||
XM_017028161.1 | Intron | XP_016883650.1 |
Set Membership: |
HapMap |