Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612828 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEBPZ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CEBPZ - CCAAT/enhancer binding protein zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005760.2 | 2823 | Silent Mutation | AGC,AGT | S,S 931 | NP_005751.2 |
CEBPZOS - CEBPZ opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322373.1 | 2823 | Intron | NP_001309302.1 | |||
NM_001322374.1 | 2823 | Intron | NP_001309303.1 | |||
NM_001322375.1 | 2823 | Intron | NP_001309304.1 | |||
NM_001322376.1 | 2823 | Intron | NP_001309305.1 | |||
XM_017003106.1 | 2823 | Intron | XP_016858595.1 | |||
XM_017003107.1 | 2823 | Intron | XP_016858596.1 | |||
XM_017003108.1 | 2823 | Intron | XP_016858597.1 | |||
XM_017003109.1 | 2823 | Intron | XP_016858598.1 |
Set Membership: |
HapMap Validated |