Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602719 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRA2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRA2B - transformer 2 beta homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243879.1 | 2325 | UTR 3 | NP_001230808.1 | |||
NM_004593.2 | 2325 | UTR 3 | NP_004584.1 | |||
XM_005247703.1 | 2325 | Intron | XP_005247760.1 | |||
XM_011513072.2 | 2325 | Intron | XP_011511374.1 | |||
XM_017007033.1 | 2325 | Intron | XP_016862522.1 |