Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612978 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBLN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CBLN3 - cerebellin 3 precursor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KHNYN - KH and NYN domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290256.1 | Intron | NP_001277185.1 | ||||
NM_001290257.1 | Intron | NP_001277186.1 | ||||
NM_015299.2 | Intron | NP_056114.1 | ||||
XM_005267474.4 | Intron | XP_005267531.1 | ||||
XM_011536590.2 | Intron | XP_011534892.1 | ||||
XM_017021114.1 | Intron | XP_016876603.1 |
NYNRIN - NYN domain and retroviral integrase containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |