Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300941 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM199X PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FAM199X - family with sequence similarity 199, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207318.3 | 16 | Intron | NP_997201.1 | |||
XM_005262079.2 | 16 | Intron | XP_005262136.1 | |||
XM_017029270.1 | 16 | UTR 5 | XP_016884759.1 |
SLC25A53 - solute carrier family 25 member 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012755.4 | 16 | Intron | NP_001012773.2 | |||
XM_005262129.4 | 16 | Intron | XP_005262186.1 | |||
XM_011530952.2 | 16 | Intron | XP_011529254.1 | |||
XM_011530953.2 | 16 | Intron | XP_011529255.1 | |||
XM_017029514.1 | 16 | Intron | XP_016885003.1 | |||
XM_017029515.1 | 16 | Intron | XP_016885004.1 | |||
XM_017029516.1 | 16 | Intron | XP_016885005.1 |