Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATTAGCAAATCAAGTCTAACAA[C/T]GTATACACAGAATTGCCCACCCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602164 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HTR4 - 5-hydroxytryptamine receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000870.6 | Intron | NP_000861.1 | ||||
NM_001040169.2 | Intron | NP_001035259.1 | ||||
NM_001040172.2 | Intron | NP_001035262.2 | ||||
NM_001040173.2 | Intron | NP_001035263.1 | ||||
NM_001286410.1 | Intron | NP_001273339.1 | ||||
NM_199453.3 | Intron | NP_955525.1 |
Set Membership: |
HapMap |