Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGTATTTGTTCCAGGAACAGGG[A/G]AGCCATATTCTAAGAGTGGAATGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601436 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMBRD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LMBRD2 - LMBR1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR580 - microRNA 580 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SKP2 - S-phase kinase-associated protein 2, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243120.1 | Intron | NP_001230049.1 | ||||
NM_005983.3 | Intron | NP_005974.2 | ||||
NM_032637.3 | Intron | NP_116026.1 | ||||
XM_011514081.2 | Intron | XP_011512383.1 | ||||
XM_011514082.2 | Intron | XP_011512384.1 | ||||
XM_011514083.2 | Intron | XP_011512385.1 | ||||
XM_017009753.1 | Intron | XP_016865242.1 |