Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 121360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBFB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CBFB - core-binding factor, beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001755.2 | Intron | NP_001746.1 | ||||
NM_022845.2 | Intron | NP_074036.1 | ||||
XM_005256212.3 | Intron | XP_005256269.1 | ||||
XM_006721321.2 | Intron | XP_006721384.1 | ||||
XM_017023812.1 | Intron | XP_016879301.1 | ||||
XM_017023813.1 | Intron | XP_016879302.1 |
Set Membership: |
HapMap |