Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAAAAAGATATGTTTTTAATTCT[A/G]TTAGGAAGAGTGATAGAACTATGCA
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 121360 | ||||||||||||||||||||||||||
Literature Links: |
CBFB PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CBFB - core-binding factor, beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001755.2 | Intron | NP_001746.1 | ||||
NM_022845.2 | Intron | NP_074036.1 | ||||
XM_005256212.3 | Intron | XP_005256269.1 | ||||
XM_006721321.2 | Intron | XP_006721384.1 | ||||
XM_017023812.1 | Intron | XP_016879301.1 | ||||
XM_017023813.1 | Intron | XP_016879302.1 |
Set Membership: |
HapMap |