Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGAGGGAAGGGAGAATGATTAA[C/T]GACCCCAGGCATAGTGGGGTGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605322 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376777 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105376777 - uncharacterized LOC105376777 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WFDC1 - WAP four-disulfide core domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282466.1 | Intron | NP_001269395.1 | ||||
NM_001282467.1 | Intron | NP_001269396.1 | ||||
NM_021197.3 | Intron | NP_067020.2 | ||||
XM_005256079.3 | Intron | XP_005256136.1 |