Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAGATACAGAAACACTTCTTT[A/T]GATGAAGGGAGCTGGCAGCAGCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL36 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHL36 - kelch like family member 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303451.1 | Intron | NP_001290380.1 | ||||
NM_024731.3 | Intron | NP_079007.2 | ||||
XM_005256149.2 | Intron | XP_005256206.1 | ||||
XM_017023674.1 | Intron | XP_016879163.1 | ||||
XM_017023675.1 | Intron | XP_016879164.1 |