Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCACTGCAACTAATTTTAGGCC[A/G]TATTTCATTCTAGTAAGGGTTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176960 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRKCA - protein kinase C alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002737.2 | Intron | NP_002728.1 | ||||
XM_017024836.1 | Intron | XP_016880325.1 | ||||
XM_017024837.1 | Intron | XP_016880326.1 | ||||
XM_017024838.1 | Intron | XP_016880327.1 | ||||
XM_017024839.1 | Intron | XP_016880328.1 | ||||
XM_017024840.1 | Intron | XP_016880329.1 | ||||
XM_017024841.1 | Intron | XP_016880330.1 |
Set Membership: |
HapMap |