Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGGCGATTGCACTGCGCGCGGT[A/C]CAAGTTGCTGGCGAGCCAGGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKCA - protein kinase C alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002737.2 | Intron | NP_002728.1 | ||||
XM_017024836.1 | Intron | XP_016880325.1 | ||||
XM_017024837.1 | Intron | XP_016880326.1 | ||||
XM_017024838.1 | Intron | XP_016880327.1 | ||||
XM_017024839.1 | Intron | XP_016880328.1 | ||||
XM_017024840.1 | Intron | XP_016880329.1 | ||||
XM_017024841.1 | Intron | XP_016880330.1 |