Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTCCCCTTCTCTCTGGTGACCT[C/T]GTGGCTTGGTACTTGCGTGGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607173 MIM: 604616 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMD14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PSMD14 - proteasome 26S subunit, non-ATPase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBR1 - T-box, brain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006593.3 | Intron | NP_006584.1 |
Set Membership: |
HapMap Validated |