Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTAAGTGTACAAAACACACAAAC[A/T]TCACTTTACTTGGAAAATTATTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601756 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GALNT3 - polypeptide N-acetylgalactosaminyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004482.3 | 2453 | UTR 3 | NP_004473.2 | |||
XM_005246449.1 | 2453 | UTR 3 | XP_005246506.1 | |||
XM_011510929.1 | 2453 | UTR 3 | XP_011509231.1 | |||
XM_017003770.1 | 2453 | UTR 3 | XP_016859259.1 |
Set Membership: |
HapMap |