Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611110 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCT8 - chaperonin containing TCP1 subunit 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP3K7CL - MAP3K7 C-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286617.1 | Intron | NP_001273546.1 | ||||
NM_001286618.1 | Intron | NP_001273547.1 | ||||
NM_001286619.1 | Intron | NP_001273548.1 | ||||
NM_001286620.1 | Intron | NP_001273549.1 | ||||
NM_001286622.1 | Intron | NP_001273551.1 | ||||
NM_001286623.1 | Intron | NP_001273552.1 | ||||
NM_001286624.1 | Intron | NP_001273553.1 | ||||
NM_001286634.1 | Intron | NP_001273563.1 | ||||
NM_020152.3 | Intron | NP_064537.1 |