Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610109 MIM: 613160 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANO2 - anoctamin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278596.1 | Intron | NP_001265525.1 | ||||
NM_001278597.1 | Intron | NP_001265526.1 | ||||
XM_006718988.3 | Intron | XP_006719051.1 | ||||
XM_011520975.2 | Intron | XP_011519277.1 | ||||
XM_011520978.2 | Intron | XP_011519280.1 | ||||
XM_017019672.1 | Intron | XP_016875161.1 |
VWF - von Willebrand factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000552.4 | Intron | NP_000543.2 |
Set Membership: |
HapMap |