Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATGATAAACTTACATGGATTCT[C/G]TATCCATAGGGAAGAAAAGGAGTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613644 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF7IP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ATF7IP - activating transcription factor 7 interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLBD1 - phospholipase B domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024829.5 | Intron | NP_079105.4 |
Set Membership: |
HapMap Validated |