Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGACTCACTGCAAGACTGACAT[A/C]TAATCCCTGGACAATAGAATGCTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF426 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF426 - zinc finger protein 426 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300883.2 | 4337 | UTR 3 | NP_001287812.1 | |||
NM_001318055.1 | 4337 | Intron | NP_001304984.1 | |||
NM_001318056.1 | 4337 | Intron | NP_001304985.1 | |||
NM_024106.2 | 4337 | Intron | NP_077011.1 | |||
XM_017027293.1 | 4337 | Intron | XP_016882782.1 | |||
XM_017027294.1 | 4337 | Intron | XP_016882783.1 |
Set Membership: |
HapMap |