Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610349 MIM: 609623 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAMSTR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAMSTR - MEF2 activating motif and SAP domain containing transcriptional regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASIP1 - Ras interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017805.2 | 3042 | Intron | NP_060275.2 | |||
XM_011527053.2 | 3042 | Intron | XP_011525355.2 | |||
XM_011527054.2 | 3042 | Missense Mutation | AGG,GGG | R,G 1010 | XP_011525356.2 | |
XM_011527055.2 | 3042 | Intron | XP_011525357.2 | |||
XM_017026914.1 | 3042 | Intron | XP_016882403.1 | |||
XM_017026915.1 | 3042 | Intron | XP_016882404.1 |