Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTTCAAGACAGTTTTTGGGGTG[C/T]GGGAAGTATCCACATTCCTACTTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142705 MIM: 603144 MIM: 606936 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HRC - histidine rich calcium binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPFIA3 - PTPRF interacting protein alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRPM4 - transient receptor potential cation channel subfamily M member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195227.1 | Intron | NP_001182156.1 | ||||
NM_001321281.1 | Intron | NP_001308210.1 | ||||
NM_001321282.1 | Intron | NP_001308211.1 | ||||
NM_001321283.1 | Intron | NP_001308212.1 | ||||
NM_001321285.1 | Intron | NP_001308214.1 | ||||
NM_017636.3 | Intron | NP_060106.2 | ||||
XM_005259017.1 | Intron | XP_005259074.1 |
Set Membership: |
HapMap |