Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608879 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS13C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VPS13C - vacuolar protein sorting 13 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018088.2 | Intron | NP_001018098.1 | ||||
NM_017684.4 | Intron | NP_060154.3 | ||||
NM_018080.3 | Intron | NP_060550.2 | ||||
NM_020821.2 | Intron | NP_065872.1 | ||||
XM_011521713.2 | Intron | XP_011520015.1 | ||||
XM_011521714.2 | Intron | XP_011520016.1 |
Set Membership: |
HapMap |