Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118495 MIM: 601441 MIM: 162096 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRM4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHRM4 - cholinergic receptor muscarinic 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000741.3 | 1302 | Silent Mutation | AAC,AAT | N,N 417 | NP_000732.2 |
DGKZ - diacylglycerol kinase zeta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MDK - midkine (neurite growth-promoting factor 2) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4688 - microRNA 4688 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |