Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCCTTAATGTTTAATTTGAACT[C/T]TTTTGTATTGTGGTCTGAAATAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602019 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375744 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105375744 - uncharacterized LOC105375744 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SQLE - squalene epoxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003129.3 | Intron | NP_003120.2 | ||||
XM_011517246.2 | Intron | XP_011515548.1 |
Set Membership: |
Validated |