Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATACCCAGAACTTATCCATCTTA[C/T]AACTGAAAGTCTGTGCCCTTTAACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604585 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SP100 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SP100 - SP100 nuclear antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080391.1 | Intron | NP_001073860.1 | ||||
NM_001206701.1 | Intron | NP_001193630.1 | ||||
NM_001206702.1 | Intron | NP_001193631.1 | ||||
NM_001206703.1 | Intron | NP_001193632.1 | ||||
NM_001206704.1 | Intron | NP_001193633.1 | ||||
NM_003113.3 | Intron | NP_003104.2 |