Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613831 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UBR3 - ubiquitin protein ligase E3 component n-recognin 3 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172070.3 | Intron | NP_742067.3 | ||||
XM_005246299.4 | Intron | XP_005246356.1 | ||||
XM_005246300.4 | Intron | XP_005246357.1 | ||||
XM_006712268.3 | Intron | XP_006712331.1 | ||||
XM_006712269.3 | Intron | XP_006712332.1 | ||||
XM_006712270.3 | Intron | XP_006712333.2 | ||||
XM_006712271.3 | Intron | XP_006712334.1 | ||||
XM_011510604.2 | Intron | XP_011508906.1 | ||||
XM_017003356.1 | Intron | XP_016858845.1 | ||||
XM_017003357.1 | Intron | XP_016858846.1 | ||||
XM_017003358.1 | Intron | XP_016858847.1 | ||||
XM_017003359.1 | Intron | XP_016858848.1 |
Set Membership: |
HapMap |