Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAACAGTGTCAAAGACTGTCAGA[C/G]TGACCTTGAAATCATTTAGCAATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608438 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TLK1 - tousled like kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136554.1 | 4790 | UTR 3 | NP_001130026.1 | |||
NM_001136555.1 | 4790 | UTR 3 | NP_001130027.1 | |||
NM_012290.4 | 4790 | UTR 3 | NP_036422.3 | |||
XM_011512238.2 | 4790 | Intron | XP_011510540.1 | |||
XM_017005421.1 | 4790 | Intron | XP_016860910.1 | |||
XM_017005422.1 | 4790 | Intron | XP_016860911.1 |