Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612991 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASXL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ASXL2 - additional sex combs like 2, transcriptional regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018263.4 | 11298 | Intron | NP_060733.4 | |||
XM_006712039.3 | 11298 | UTR 3 | XP_006712102.1 | |||
XM_006712040.2 | 11298 | UTR 3 | XP_006712103.1 | |||
XM_011532950.2 | 11298 | UTR 3 | XP_011531252.1 | |||
XM_011532951.2 | 11298 | UTR 3 | XP_011531253.1 | |||
XM_017004429.1 | 11298 | UTR 3 | XP_016859918.1 | |||
XM_017004430.1 | 11298 | UTR 3 | XP_016859919.1 |